3-Iodoaniline 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR4934-5G
OR4934-10G
OR4934-25G
APOSOR4934-5GEA
23.4
EUR
APOSOR4934-5G
APOSOR4934-10G
APOSOR4934-25G
3-Iodoaniline 98%
3-Iodoaniline
Formule:
C₆H₆IN Kookpunt: 145…146 °C/15mmHg Smeltpunt: 21…24 °C Dichtheid: 1,821 g/cm³ (25 °C) Vlampunt: 113 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00007781 CAS nummer: 626-01-7 EINECS: 210-922-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...