3-Thienylacetic acid 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Thiophene-3-acetic acid, 3-Thiopheneacetic acid
OR5060-25G
OR5060-5G
OR5060-100G
APOSOR5060-25GEA
42.4
EUR
APOSOR5060-25G
APOSOR5060-5G
APOSOR5060-100G
3-Thienylacetic acid 98%
3-Thienylacetic acid
Formule:
C₆H₆O₂S Smeltpunt: 75…77 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005473 CAS nummer: 6964-21-2 EINECS: 230-166-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...