2-Ethoxyacetyl chloride 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR50944-100G
OR50944-25G
OR50944-5G
APOSOR50944-100GEA
172
EUR
APOSOR50944-100G
APOSOR50944-25G
APOSOR50944-5G
2-Ethoxyacetyl chloride 97%
2-Ethoxyacetyl chloride
Formule:
C₄H₇ClO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD11170566 CAS nummer: 14077-58-8 UN: 2920 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...