Cyclohexanemethylamine 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
(Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine
OR5167-25G
OR5167-100G
APOSOR5167-25GEA
191
EUR
APOSOR5167-25G
APOSOR5167-100G
Cyclohexanemethylamine 95%
Cyclohexanemethylamine
Formule:
C₇H₁₅N MW: 113.2 g/mol Kookpunt: 159…163 °C Smeltpunt: –8 °C Dichtheid: 0.871 g/cm³ (25 °C) Vlampunt: 43 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001520 CAS nummer: 3218-02-8 EINECS: 221-741-8 UN: 2734 ADR: 8+3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...