N'-Hydroxycyclohexanecarboximidamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR52382-1G OR52382-250MG
APOSOR52382-1GEA 371 EUR
APOSOR52382-1G APOSOR52382-250MG
N'-Hydroxycyclohexanecarboximidamide
N'-Hydroxycyclohexanecarboximidamide
Formule: C₇H₁₄N₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07776175
CAS nummer: 633313-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR