1-Decanethiol 96%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan
OR5323-100ML
OR5323-2.5L
OR5323-500ML
APOSOR5323-100MLEA
35.1
EUR
APOSOR5323-100ML
APOSOR5323-2.5L
APOSOR5323-500ML
1-Decanethiol 96%
1-Decanethiol
Formule:
C₁₀H₂₂S Kookpunt: 114 °C/13mmHg Smeltpunt: –26 °C Dichtheid: 0,843 g/cm³ (25 °C) Vlampunt: 98 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00004884 CAS nummer: 143-10-2 EINECS: 205-584-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...