4-Benzyloxybenzyl alcohol 95+%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(4-(Benzyloxy)phenyl)methanol
OR5343-100G
OR5343-25G
APOSOR5343-100GEA
175
EUR
APOSOR5343-100G
APOSOR5343-25G
4-Benzyloxybenzyl alcohol 95+%
4-Benzyloxybenzyl alcohol
Formule:
C₁₄H₁₄O₂ Smeltpunt: 86…87 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004654 CAS nummer: 836-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...