3-Thiopheneboronic acid
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR5445-25G
OR5445-5G
APOSOR5445-25GEA
0
EUR
APOSOR5445-25G
APOSOR5445-5G
3-Thiopheneboronic acid
3-Thiopheneboronic acid
Formule:
C₄H₅BO₂S Smeltpunt: 164…169 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00151851 CAS nummer: 6165-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...