(S)-(+)-Mandelic acid 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
APOSOR5460-1KGEA
104
EUR
APOSOR5460-1KG
(S)-(+)-Mandelic acid 97%
(S)-(+)-Mandelic acid
Formule:
C₈H₈O₃ MW: 152,15 g/mol Smeltpunt: 131…134 °C Vlampunt: 200 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004495 CAS nummer: 17199-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...