Monosodium 5-sulphoisophthalate 98+%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Sodium 3,5-dicarboxybenzenesulphonate, 5-Sulphoisophthalic acid monosodium salt, Sodium 5-sulphoisophthalate
OR54628-25G
OR54628-100G
APOSOR54628-25GEA
49.8
EUR
APOSOR54628-25G
APOSOR54628-100G
Monosodium 5-sulphoisophthalate 98+%
Monosodium 5-sulphoisophthalate
Formule:
C₈H₆O₇S Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007495 CAS nummer: 6362-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...