2-Amino-4-(4-bromophenyl)thiazole 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR5603-25G
OR5603-5G
OR5603-1G
APOSOR5603-25GEA
87.6
EUR
APOSOR5603-25G
APOSOR5603-5G
APOSOR5603-1G
2-Amino-4-(4-bromophenyl)thiazole 98%
2-Amino-4-(4-bromophenyl)thiazole
Formule:
C₉H₇BrN₂S MW: 255.1343 g/mol Smeltpunt: 181…187 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00051953 CAS nummer: 2103-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...