3-Mercaptohexyl acetate 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Acetic Acid 3-Mercaptohexyl Ester
OR5751-1G
OR5751-5G
OR5751-10G
APOSOR5751-1GEA
52.6
EUR
APOSOR5751-1G
APOSOR5751-5G
APOSOR5751-10G
3-Mercaptohexyl acetate 95%
3-Mercaptohexyl acetate
Formule:
C₈H₁₆O₂S Kookpunt: 240 °C/570mmHg Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00792516 CAS nummer: 136954-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...