Di-tert-butyl azodicarboxylate 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Di-tert-butyl diazene-1,2-dicarboxylate
OR5753-100G OR5753-25G OR5753-5G
APOSOR5753-100GEA 52.6 EUR
APOSOR5753-100G APOSOR5753-25G APOSOR5753-5G
Di-tert-butyl azodicarboxylate 98%
Di-tert-butyl azodicarboxylate
Formule: C₁₀H₁₈N₂O₄
MW: 230.26 g/mol
Smeltpunt: 89…92 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00015001
CAS nummer: 870-50-8
EINECS: 212-769-9