N-Benzyl-3-bromobenzenesulphonamide 96%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR5809-1G OR5809-25G OR5809-5G
APOSOR5809-1GEA 107 EUR
APOSOR5809-1G APOSOR5809-25G APOSOR5809-5G
N-Benzyl-3-bromobenzenesulphonamide 96%
N-Benzyl-3-bromobenzenesulphonamide
Formule: C₁₃H₁₂BrNO₂S
Smeltpunt: 78…82 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07363821
CAS nummer: 625470-36-2
Order Now
Specificaties
Environmentally Preferable