3-Chloropropanesulphonyl chloride 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR5825-100G OR5825-25G OR5825-5G
APOSOR5825-100GEA 193 EUR
APOSOR5825-100G APOSOR5825-25G APOSOR5825-5G
3-Chloropropanesulphonyl chloride 98%
3-Chloropropanesulphonyl chloride
Formule: C₃H₆Cl₂O₂S
Kookpunt: 70 °C/0,5mmHg
Dichtheid: 1,456 g/cm³ (25 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007463
CAS nummer: 1633-82-5
EINECS: 216-646-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR