5-Bromo-o-toluic acid
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR59443-25G
OR59443-100G
APOSOR59443-25GEA
0
EUR
APOSOR59443-25G
APOSOR59443-100G
5-Bromo-o-toluic acid
5-Bromo-o-toluic acid
Formule:
C₈H₇BrO₂ Smeltpunt: 167…171 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00267350 CAS nummer: 79669-49-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...