2-Bromofluorene
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR5981-100G
OR5981-25G
OR5981-5G
APOSOR5981-100GEA
48.3
EUR
APOSOR5981-100G
APOSOR5981-25G
APOSOR5981-5G
2-Bromofluorene
2-Bromofluorene
Formule:
C₁₃H₉Br MW: 245.12 g/mol Kookpunt: 185 °C/135mmHg Smeltpunt: 110…114 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001115 CAS nummer: 1133-80-8 EINECS: 214-480-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...