1,6-Naphthyridine-2-carboxylic acid 95+%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR5998-1G OR5998-250MG
APOSOR5998-1GEA 220 EUR
APOSOR5998-1G APOSOR5998-250MG
1,6-Naphthyridine-2-carboxylic acid 95+%
1,6-Naphthyridine-2-carboxylic acid
Formule: C₉H₆N₂O₂
MDL Nummer: MFCD00202908
CAS nummer: 197507-59-8