6-Isoquinolineboronic acid 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR60071-1G
OR60071-250MG
APOSOR60071-1GEA
238
EUR
APOSOR60071-1G
APOSOR60071-250MG
6-Isoquinolineboronic acid 95%
6-Isoquinolineboronic acid
Formule:
C₉H₈BNO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD10697418 CAS nummer: 899438-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...