Cyclopentylacetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Cyclopentylacetic acid

OR6624-100G OR6624-25G OR6624-5G
APOSOR6624-100GEA 277 EUR
APOSOR6624-100G APOSOR6624-25G APOSOR6624-5G
Cyclopentylacetic acid
Cyclopentylacetic acid
Formule: C₇H₁₂O₂
MW: 128.17 g/mol
Kookpunt: 133…134 °C/23mmHg
Dichtheid: 1.023 g/cm³ (25 °C)
Vlampunt: 109 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001387
CAS nummer: 1123-00-8
EINECS: 214-368-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR