Cyclopentanecarbonyl chloride 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Cyclopentanecarboxylic chloride
OR6700-100G
OR6700-25G
OR6700-5G
APOSOR6700-100GEA
382
EUR
APOSOR6700-100G
APOSOR6700-25G
APOSOR6700-5G
Cyclopentanecarbonyl chloride 98%
Cyclopentanecarbonyl chloride
Formule:
C₆H₉ClO MW: 132.59 g/mol Kookpunt: 161…162 °C Dichtheid: 1.091 g/cm³ (25 °C) Vlampunt: 60 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001370 CAS nummer: 4524-93-0 EINECS: 224-856-1 UN: 2920 ADR: 8+3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...