3-Indazolinone 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR7741-100G
OR7741-25G
OR7741-5G
APOSOR7741-100GEA
600
EUR
APOSOR7741-100G
APOSOR7741-25G
APOSOR7741-5G
3-Indazolinone 97%
3-Indazolinone
Formule:
C₇H₆N₂O Smeltpunt: 224…226 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005685 CAS nummer: 7364-25-2 EINECS: 230-904-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...