3-(2,3-Dihydrobenzofuran-5-yl)propanoic acid 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,3-Dihydro-1-benzofuran-5-propanoic acid

OR7838-1G OR7838-250MG
APOSOR7838-1GEA 0 EUR
APOSOR7838-1G APOSOR7838-250MG
3-(2,3-Dihydrobenzofuran-5-yl)propanoic acid 95+%
3-(2,3-Dihydrobenzofuran-5-yl)propanoic acid
Formule: C₁₁H₁₂O₃
Smeltpunt: 144…148 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03102298
CAS nummer: 215057-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR