5-Bromo-2-hydrazinopyrimidine
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR7905-1G
OR7905-5G
APOSOR7905-1GEA
22
EUR
APOSOR7905-1G
APOSOR7905-5G
5-Bromo-2-hydrazinopyrimidine
5-Bromo-2-hydrazinopyrimidine
Formule:
C₄H₅BrN₄ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD03452780 CAS nummer: 823-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...