Ethylboronic acid
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR8874-1G
OR8874-5G
APOSOR8874-1GEA
22
EUR
APOSOR8874-1G
APOSOR8874-5G
Ethylboronic acid
Ethylboronic acid
Formule:
C₂H₇BO₂ Smeltpunt: 84…88 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD01074536 CAS nummer: 4433-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...