2-Chloro-3-(chloromethyl)pyrazine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR903175-1G OR903175-250MG
APOSOR903175-1GEA 651 EUR
APOSOR903175-1G APOSOR903175-250MG
2-Chloro-3-(chloromethyl)pyrazine 95%
2-Chloro-3-(chloromethyl)pyrazine
Formule: C₅H₄Cl₂N₂
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 45660-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR