Pyrazinamide 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide
OR9043-100G
OR9043-500G
APOSOR9043-100GEA
130
EUR
APOSOR9043-100G
APOSOR9043-500G
Pyrazinamide 98%
Pyrazinamide
Formule:
C₅H₅N₃O Smeltpunt: 189…191 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006132 CAS nummer: 98-96-4 EINECS: 202-717-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...