2-Pyrimidinecarbonitrile 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR9045-100G
OR9045-500G
OR9045-1G
APOSOR9045-100GEA
0
EUR
APOSOR9045-100G
APOSOR9045-500G
APOSOR9045-1G
2-Pyrimidinecarbonitrile 98%
2-Pyrimidinecarbonitrile
Formule:
C₅H₃N₃ Smeltpunt: 37…44 °C Vlampunt: >110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00160513 CAS nummer: 14080-23-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...