N,N,N-Trimethylanilinium tribromide 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide
OR904742-100G
OR904742-1KG
OR904742-500G
APOSOR904742-100GEA
0
EUR
APOSOR904742-100G
APOSOR904742-1KG
APOSOR904742-500G
N,N,N-Trimethylanilinium tribromide 98%
N,N,N-Trimethylanilinium tribromide
Formule:
C₉H₁₄Br₃N MW: 375.93 g/mol Smeltpunt: 112…117 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011789 CAS nummer: 4207-56-1 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...