(4-Acetamidophenyl)boronic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Acetamidophenylboronic acid, 4-Acetamidobenzeneboronic acid, 4-Acetylaminophenylboronic acid

OR9048-5G OR9048-25G
APOSOR9048-5GEA 93.5 EUR
APOSOR9048-5G APOSOR9048-25G
(4-Acetamidophenyl)boronic acid
(4-Acetamidophenyl)boronic acid
Formule: C₈H₁₀BNO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02179451
CAS nummer: 101251-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR