5-Chloro-8-methylquinoline 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR909168-1G
OR909168-5G
APOSOR909168-1GEA
106
EUR
APOSOR909168-1G
APOSOR909168-5G
5-Chloro-8-methylquinoline 98%
5-Chloro-8-methylquinoline
Formule:
C₁₀H₈ClN Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00463515 CAS nummer: 78941-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...