Triphenylethylene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,1,2-Triphenylethylene, 1,1,2-Triphenylethene

OR9099-10G OR9099-250G OR9099-50G
APOSOR9099-10GEA 22 EUR
APOSOR9099-10G APOSOR9099-250G APOSOR9099-50G
Triphenylethylene
Triphenylethylene
Formule: (C₆H₅)₂CCHC₆H₅
Kookpunt: 219…221 °C/14mmHg
Smeltpunt: 67…71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004765
CAS nummer: 58-72-0
EINECS: 200-395-1
UN: 3077
ADR: 9+EHS,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR