3-Bromo-7-nitroindole 96%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR912799-1G
OR912799-25G
OR912799-5G
APOSOR912799-1GEA
76.4
EUR
APOSOR912799-1G
APOSOR912799-25G
APOSOR912799-5G
3-Bromo-7-nitroindole 96%
3-Bromo-7-nitroindole
Formule:
C₈H₅BrN₂O₂ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD02684155 CAS nummer: 397864-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...