5-Isopropylisoxazole-3-carboxylic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR913177-10G OR913177-1G OR913177-5G
APOSOR913177-10GEA 388 EUR
APOSOR913177-10G APOSOR913177-1G APOSOR913177-5G
5-Isopropylisoxazole-3-carboxylic acid 97%
5-Isopropylisoxazole-3-carboxylic acid
Formule: C₇H₉NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06738923
CAS nummer: 89776-74-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR