(S)-(-)-Bromosuccinic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR913781-1G OR913781-5G
APOSOR913781-1GEA 96.4 EUR
APOSOR913781-1G APOSOR913781-5G
(S)-(-)-Bromosuccinic acid 95%
(S)-(-)-Bromosuccinic acid
Formule: HOOCBrCHCH₂COOH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00153931
CAS nummer: 20859-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR