(S)-(+)-Ketopinic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR913787-1G OR913787-5G
APOSOR913787-1GEA 33.2 EUR
APOSOR913787-1G APOSOR913787-5G
(S)-(+)-Ketopinic acid 95%
(S)-(+)-Ketopinic acid
Formule: C₁₀H₁₄O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00168053
CAS nummer: 40724-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR