Isopropyl carbamate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR916469-100G OR916469-25G OR916469-5G
APOSOR916469-100GEA 347 EUR
APOSOR916469-100G APOSOR916469-25G APOSOR916469-5G
Isopropyl carbamate 95%
Isopropyl carbamate
Formule: C₄H₉NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00025463
CAS nummer: 1746-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR