Ethyl phenethylcarbamate 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR917901-1G
OR917901-250MG
APOSOR917901-1GEA
157
EUR
APOSOR917901-1G
APOSOR917901-250MG
Ethyl phenethylcarbamate 95%
Ethyl phenethylcarbamate
Formule:
C₁₁H₁₅NO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00026817 CAS nummer: 6970-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...