2-Pentyl-2-cyclopenten-1-one 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR918346-25G
OR918346-5G
APOSOR918346-25GEA
315
EUR
APOSOR918346-25G
APOSOR918346-5G
2-Pentyl-2-cyclopenten-1-one 95%
2-Pentyl-2-cyclopenten-1-one
Formule:
C₁₀H₁₆O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00036631 CAS nummer: 25564-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...