2-Oxoindoline-4-carboxylic acid 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR929067-1G
OR929067-250MG
APOSOR929067-1GEA
122
EUR
APOSOR929067-1G
APOSOR929067-250MG
2-Oxoindoline-4-carboxylic acid 95%
2-Oxoindoline-4-carboxylic acid
Formule:
C₉H₇NO₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD10000702 CAS nummer: 90322-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...