5-Butylpicolinic acid 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR933247-1G
OR933247-250MG
APOSOR933247-1GEA
548
EUR
APOSOR933247-1G
APOSOR933247-250MG
5-Butylpicolinic acid 95%
5-Butylpicolinic acid
Formule:
C₁₀H₁₃NO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006298 CAS nummer: 536-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...