6-Acetyl-2(3H)-benzothiazolone 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR936467-1G OR936467-5G
APOSOR936467-1GEA 132 EUR
APOSOR936467-1G APOSOR936467-5G
6-Acetyl-2(3H)-benzothiazolone 95%
6-Acetyl-2(3H)-benzothiazolone
Formule: C₉H₇NO₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02660572
CAS nummer: 133044-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR