3-(2-Furyl)-2-phenylpropenal 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR936490-1G OR936490-25G OR936490-5G
APOSOR936490-1GEA 132 EUR
APOSOR936490-1G APOSOR936490-25G APOSOR936490-5G
3-(2-Furyl)-2-phenylpropenal 95%
3-(2-Furyl)-2-phenylpropenal
Formule: C₁₃H₁₀O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036566
CAS nummer: 57568-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR