4-Aminostilbene 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR936508-1G
OR936508-5G
APOSOR936508-1GEA
111
EUR
APOSOR936508-1G
APOSOR936508-5G
4-Aminostilbene 95%
4-Aminostilbene
Formule:
C₁₄H₁₃N Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00025377 CAS nummer: 834-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...