(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR939188-1G OR939188-250MG
APOSOR939188-1GEA 409 EUR
APOSOR939188-1G APOSOR939188-250MG
(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene 95%
(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene
Formule: C₁₀H₁₄O
MW: 150.22 g/mol
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 911200-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR