Sorbic chloride 80%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR939977-1G OR939977-5G
APOSOR939977-1GEA 89.3 EUR
APOSOR939977-1G APOSOR939977-5G
Sorbic chloride 80%
Sorbic chloride
Formule: C₆H₇ClO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00045227
CAS nummer: 2614-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR