1-Ethylcyclohexanol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR941141-1G OR941141-5G
APOSOR941141-1GEA 21.7 EUR
APOSOR941141-1G APOSOR941141-5G
1-Ethylcyclohexanol 95%
1-Ethylcyclohexanol
Formule: C₈H₁₆O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00021402
CAS nummer: 1940-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR