Methyl-2-oxotetrahydrofuran-3-carboxylate 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR950450-1G
OR950450-250MG
APOSOR950450-1GEA
64.3
EUR
APOSOR950450-1G
APOSOR950450-250MG
Methyl-2-oxotetrahydrofuran-3-carboxylate 95%
Methyl-2-oxotetrahydrofuran-3-carboxylate
Formule:
C₆H₈O₄ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD10569624 CAS nummer: 19406-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...