4-Methylphenethyl bromide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR951094-1G OR951094-5G
APOSOR951094-1GEA 21.7 EUR
APOSOR951094-1G APOSOR951094-5G
4-Methylphenethyl bromide 97%
4-Methylphenethyl bromide
Formule: C₉H₁₁Br
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00050368
CAS nummer: 6529-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR