2-Bromo-1-cyclopropylethanone 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR951324-25G OR951324-5G
APOSOR951324-25GEA 83.3 EUR
APOSOR951324-25G APOSOR951324-5G
2-Bromo-1-cyclopropylethanone 97%
2-Bromo-1-cyclopropylethanone
Formule: C₅H₇BrO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08460238
CAS nummer: 69267-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR